Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3beta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202127.2 | NP_001189056.1 | 1 - 2 | 370 | 76 | |
NM_001202128.2 | NP_001189057.1 | 1 - 2 | 370 | 76 | ||
NM_139855.4 | NP_648112.2 | 1 - 2 | 370 | 76 | ||
NM_168210.3 | NP_729248.1 | 2 - 3 | 274 | 76 | ||
GenBank mRNA | AY061162.1 | - | 1 - 2 | 327 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |