Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274617.1 | NP_001261546.1 | 1 - 2 | 2370 | 89 | |
NM_168236.3 | NP_729300.1 | 1 - 2 | 2370 | 89 | ||
GenBank mRNA | AY775786.1 | - | 1 - 2 | 204 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |