Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139967.2 | NP_648224.1 | 1 - 2 | 912 | 68 | |
NM_168276.2 | NP_729382.1 | 1 - 2 | 912 | 68 | ||
GenBank mRNA | AY118593.1 | - | 1 - 2 | 577 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |