Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169916.2 | NP_001163387.2 | 7 - 8 | 4284 | 92 | |
NM_001202142.2 | NP_001189071.1 | 5 - 6 | 867 | 92 | ||
NM_001202143.2 | NP_001189072.1 | 5 - 6 | 867 | 92 | ||
NM_001259753.2 | NP_001246682.1 | 4 - 5 | 546 | 92 | ||
NM_001259754.2 | NP_001246683.1 | 5 - 6 | 867 | 92 | ||
NM_001259755.1 | NP_001246684.1 | 5 - 6 | 1869 | 92 | ||
NM_001274670.1 | NP_001261599.1 | 7 - 8 | 4808 | 92 | ||
NM_168293.3 | NP_729410.1 | 5 - 6 | 867 | 92 | ||
NM_168294.3 | NP_729411.1 | 5 - 6 | 867 | 92 | ||
GenBank mRNA | BT050456.1 | - | 5 - 6 | 833 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |