Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259770.1 | NP_001246699.1 | 26 - 27 | 10457 | 87 | |
NM_001259771.1 | NP_001246700.1 | 25 - 26 | 9954 | 87 | ||
NM_001259772.1 | NP_001246701.1 | 26 - 27 | 10116 | 87 | ||
NM_001274708.1 | NP_001261637.1 | 24 - 25 | 10283 | 87 | ||
NM_001274709.1 | NP_001261638.1 | 26 - 27 | 10098 | 87 | ||
NM_001274710.1 | NP_001261639.1 | 25 - 26 | 9972 | 87 | ||
NM_168356.2 | NP_729519.2 | 25 - 26 | 10068 | 87 | ||
NM_168357.3 | NP_729520.1 | 24 - 25 | 9924 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |