Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259770.1 | NP_001246699.1 | 3 - 4 | 2628 | 63 | |
NM_001259771.1 | NP_001246700.1 | 3 - 4 | 2269 | 63 | ||
NM_001259772.1 | NP_001246701.1 | 3 - 4 | 2269 | 63 | ||
NM_001274708.1 | NP_001261637.1 | 3 - 4 | 2628 | 63 | ||
NM_001274709.1 | NP_001261638.1 | 3 - 4 | 2269 | 63 | ||
NM_001274710.1 | NP_001261639.1 | 3 - 4 | 2269 | 63 | ||
NM_168356.2 | NP_729519.2 | 3 - 4 | 2269 | 63 | ||
NM_168357.3 | NP_729520.1 | 3 - 4 | 2269 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |