Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104107.2 | NP_001097577.1 | 4 - 5 | 499 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |