Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104139.2 | NP_001097609.1 | 5 - 6 | 1364 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |