Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259854.2 | NP_001246783.1 | 11 - 12 | 3479 | 59 | |
NM_001259855.2 | NP_001246784.1 | 12 - 13 | 3379 | 59 | ||
NM_001259856.2 | NP_001246785.1 | 12 - 13 | 3635 | 59 | ||
NM_001259857.2 | NP_001246786.1 | 12 - 13 | 3382 | 59 | ||
NM_001274990.1 | NP_001261919.1 | 10 - 11 | 2842 | 59 | ||
NM_001382059.1 | NP_001369057.1 | 10 - 11 | 2842 | 59 | ||
NM_168646.3 | NP_730099.2 | 11 - 12 | 2964 | 59 | ||
NM_168648.3 | NP_730101.2 | 10 - 11 | 2817 | 59 | ||
GenBank mRNA | AF500094.1 | - | 9 - 10 | 2349 | 59 | |
BT133053.1 | - | 11 - 12 | 2952 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |