Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nc73EF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104159.3 | NP_001097629.1 | 5 - 6 | 628 | 65 | |
NM_168701.5 | NP_730223.1 | 5 - 6 | 628 | 65 | ||
NM_168702.2 | NP_730224.1 | 5 - 6 | 589 | 65 | ||
NM_168703.5 | NP_730225.1 | 5 - 6 | 631 | 65 | ||
NM_168704.4 | NP_730226.3 | 6 - 7 | 976 | 65 | ||
NM_176341.3 | NP_788519.1 | 5 - 6 | 582 | 65 | ||
GenBank mRNA | BT003743.1 | - | 5 - 6 | 609 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |