Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ome | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104136.2 | NP_001097606.1 | 7 - 8 | 2645 | 72 | |
NM_001144483.2 | NP_001137955.1 | 6 - 7 | 2684 | 72 | ||
NM_001274915.1 | NP_001261844.1 | 5 - 6 | 1838 | 72 | ||
NM_080000.4 | NP_524739.1 | 6 - 7 | 1931 | 72 | ||
GenBank mRNA | AY058514.1 | - | 6 - 7 | 1904 | 72 | |
BT024224.1 | - | 6 - 7 | 2655 | 72 | ||
BT029957.1 | - | 7 - 8 | 2629 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |