Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258636.2 | NP_001245565.1 | 3 - 4 | 1355 | 64 | |
NM_001272381.2 | NP_001259310.1 | 2 - 3 | 2315 | 64 | ||
NM_080025.2 | NP_524764.1 | 2 - 3 | 1595 | 64 | ||
GenBank mRNA | X07985.1 | - | 2 - 3 | 1595 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |