Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ergic53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080037.3 | NP_524776.1 | 2 - 3 | 932 | 81 | |
NM_176301.2 | NP_788479.1 | 2 - 3 | 932 | 81 | ||
GenBank mRNA | AF223385.1 | - | 2 - 3 | 922 | 81 | |
AY051971.1 | - | 2 - 3 | 922 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |