Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275100.1 | NP_001262029.1 | 1 - 2 | 1144 | 60 | |
NM_001275101.1 | NP_001262030.1 | 1 - 2 | 1144 | 60 | ||
NM_080049.3 | NP_524788.2 | 1 - 2 | 1144 | 60 | ||
GenBank mRNA | BT015258.1 | - | 1 - 2 | 1109 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |