Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fend | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272443.2 | NP_001259372.1 | 1 - 2 | 1158 | 78 | |
NM_132300.3 | NP_572528.1 | 1 - 2 | 1099 | 78 | ||
GenBank mRNA | AB065095.1 | - | 1 - 2 | 1078 | 78 | |
AF209858.1 | - | 1 - 2 | 1077 | 78 | ||
BT030985.1 | - | 1 - 2 | 655 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |