Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fend | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272443.2 | NP_001259372.1 | 2 - 3 | 1208 | 86 | |
NM_132300.3 | NP_572528.1 | 2 - 3 | 1149 | 86 | ||
GenBank mRNA | AB065095.1 | - | 2 - 3 | 1128 | 86 | |
AF209858.1 | - | 2 - 3 | 1127 | 86 | ||
BT030985.1 | - | 2 - 3 | 705 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |