Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132304.3 | NP_572532.1 | 1 - 2 | 787 | 79 | |
GenBank mRNA | AY051889.1 | - | 1 - 2 | 738 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |