Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cht6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103464.3 | NP_001096934.2 | 8 - 9 | 2840 | 53 | |
NM_001258669.2 | NP_001245598.1 | 9 - 10 | 2819 | 53 | ||
NM_001258670.2 | NP_001245599.1 | 8 - 9 | 2840 | 53 | ||
NM_001258671.2 | NP_001245600.1 | 8 - 9 | 2840 | 53 | ||
NM_001258672.2 | NP_001245601.1 | 8 - 9 | 2840 | 53 | ||
NM_001258673.2 | NP_001245602.1 | 9 - 10 | 2896 | 53 | ||
NM_132370.4 | NP_572598.3 | 8 - 9 | 2840 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |