Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272477.1 | NP_001259406.1 | 1 - 2 | 200 | 69 | |
NM_001298154.1 | NP_001285083.1 | 1 - 2 | 912 | 69 | ||
NM_132396.2 | NP_572624.1 | 1 - 2 | 200 | 69 | ||
GenBank mRNA | AY089361.1 | - | 1 - 2 | 185 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |