Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrac-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298219.1 | NP_001285148.1 | 1 | 558 | 89 | |
NM_132548.3 | NP_572776.1 | 1 | 558 | 89 | ||
GenBank mRNA | AY113515.2 | - | 1 | 553 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |