Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103493.4 | NP_001096963.1 | 5 - 6 | 4617 | 105 | |
NM_001103494.2 | NP_001096964.1 | 5 - 6 | 4736 | 105 | ||
NM_001298228.1 | NP_001285157.1 | 5 - 6 | 4736 | 105 | ||
NM_001298229.1 | NP_001285158.1 | 6 - 7 | 5051 | 105 | ||
NM_001298230.1 | NP_001285159.1 | 6 - 7 | 5051 | 105 | ||
NM_132584.3 | NP_572812.3 | 6 - 7 | 4925 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |