Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103495.5 | NP_001096965.1 | 3 - 4 | 1523 | 79 | |
NM_001258717.1 | NP_001245646.1 | 4 - 5 | 3043 | 79 | ||
NM_001258719.2 | NP_001245648.1 | 3 - 4 | 2609 | 79 | ||
NM_001272568.2 | NP_001259497.1 | 2 - 3 | 1315 | 79 | ||
NM_001272569.1 | NP_001259498.1 | 3 - 4 | 1781 | 79 | ||
NM_132614.6 | NP_572842.1 | 2 - 3 | 1864 | 79 | ||
GenBank mRNA | BT120384.2 | - | 1 - 2 | 200 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |