Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272584.1 | NP_001259513.1 | 4 - 5 | 1862 | 74 | |
NM_132650.2 | NP_572878.1 | 4 - 5 | 1865 | 74 | ||
GenBank mRNA | BT133177.1 | - | 4 - 5 | 1863 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |