Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yippee | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132654.4 | NP_572882.1 | 3 - 4 | 622 | 77 | |
GenBank mRNA | AF172939.1 | - | 3 - 4 | 561 | 77 | |
BT120317.1 | - | 3 - 4 | 440 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |