Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Set2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169269.2 | NP_001162740.1 | 3 - 4 | 6783 | 103 | |
NM_132660.4 | NP_572888.2 | 2 - 3 | 6994 | 103 | ||
GenBank mRNA | BT025042.1 | - | 3 - 4 | 6768 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |