Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272608.2 | NP_001259537.2 | 3 - 4 | 999 | 66 | |
NM_132700.4 | NP_572928.1 | 3 - 4 | 999 | 66 | ||
GenBank mRNA | AY058721.1 | - | 3 - 4 | 1016 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |