Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132723.3 | NP_572951.1 | 2 - 3 | 1025 | 66 | |
GenBank mRNA | AY069093.1 | - | 2 - 3 | 1004 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |