Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132723.3 | NP_572951.1 | 5 - 6 | 1949 | 106 | |
GenBank mRNA | AY069093.1 | - | 5 - 6 | 1928 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |