Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3-GEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103513.3 | NP_001096983.2 | 18 - 19 | 5646 | 99 | |
NM_001258746.2 | NP_001245675.1 | 19 - 20 | 5664 | 99 | ||
NM_001258747.1 | NP_001245676.1 | 18 - 19 | 5535 | 99 | ||
NM_001272630.1 | NP_001259559.1 | 17 - 18 | 5526 | 99 | ||
NM_001272631.1 | NP_001259560.1 | 18 - 19 | 5559 | 99 | ||
NM_001316635.1 | NP_001303564.1 | 18 - 19 | 5559 | 99 | ||
NM_001347809.1 | NP_001334679.1 | 20 - 21 | 5619 | 99 | ||
NM_001347810.1 | NP_001334680.1 | 21 - 22 | 5715 | 99 | ||
NM_001347811.1 | NP_001334681.1 | 21 - 22 | 5715 | 99 | ||
NM_132773.3 | NP_573001.2 | 17 - 18 | 5526 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |