Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272632.2 | NP_001259561.1 | 2 - 3 | 249 | 94 | |
NM_132777.3 | NP_573005.1 | 2 - 3 | 249 | 94 | ||
GenBank mRNA | BT031045.1 | - | 2 - 3 | 245 | 94 | |
KX531893.1 | - | 1 - 2 | 136 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Endogenous Controls Recommended Assay Drosophila melanogaster |