Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132874.2 | NP_573102.1 | 4 - 5 | 1048 | 61 | |
GenBank mRNA | BT003521.1 | - | 4 - 5 | 1045 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |