Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014749.2 | NP_001014749.1 | 4 - 5 | 1350 | 82 | |
NM_001103541.3 | NP_001097011.1 | 4 - 5 | 1350 | 82 | ||
NM_001272717.1 | NP_001259646.1 | 5 - 6 | 1275 | 82 | ||
NM_132992.3 | NP_573220.2 | 5 - 6 | 1275 | 82 | ||
GenBank mRNA | AY226550.1 | - | 4 - 5 | 1257 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |