Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014557.2 | NP_001014557.3 | 4 - 5 | 1841 | 72 | |
NM_001169873.1 | NP_001163344.1 | 4 - 5 | 1841 | 72 | ||
NM_001274478.1 | NP_001261407.1 | 4 - 5 | 1841 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |