Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LeuRS-m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139675.3 | NP_647932.1 | 3 - 4 | 2227 | 99 | |
GenBank mRNA | AY089557.1 | - | 3 - 4 | 2183 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |