Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir64a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139705.1 | NP_647962.1 | 6 - 7 | 2337 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |