Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139868.3 | NP_648125.3 | 3 - 4 | 1054 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |