Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139868.3 | NP_648125.3 | 4 - 5 | 1328 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |