Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043128.2 | NP_001036593.1 | 4 - 5 | 1280 | 65 | |
NM_001169901.1 | NP_001163372.1 | 5 - 6 | 1418 | 65 | ||
NM_001274610.1 | NP_001261539.1 | 5 - 6 | 1774 | 65 | ||
GenBank mRNA | BT132928.1 | - | 4 - 5 | 1100 | 65 | |
BT133054.1 | - | 5 - 6 | 1776 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |