Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139965.3 | NP_648222.3 | 2 - 3 | 319 | 85 | |
GenBank mRNA | BT133119.1 | - | 2 - 3 | 249 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |