Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259745.2 | NP_001246674.1 | 13 - 14 | 6954 | 71 | |
NM_001274657.1 | NP_001261586.1 | 12 - 13 | 7307 | 71 | ||
NM_001274658.1 | NP_001261587.1 | 13 - 14 | 6891 | 71 | ||
NM_001274659.1 | NP_001261588.1 | 13 - 14 | 7076 | 71 | ||
NM_001274660.1 | NP_001261589.1 | 14 - 15 | 7092 | 71 | ||
NM_139981.4 | NP_648238.1 | 12 - 13 | 6779 | 71 | ||
GenBank mRNA | AF299248.1 | - | 11 - 12 | 6726 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |