Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169481.2 | NP_001162952.1 | 4 - 5 | 1220 | 62 | |
NM_001273449.1 | NP_001260378.1 | 3 - 4 | 1143 | 62 | ||
NM_001273450.1 | NP_001260379.1 | 2 - 3 | 870 | 62 | ||
NM_057214.5 | NP_476562.1 | 4 - 5 | 1242 | 62 | ||
NM_057215.5 | NP_476563.1 | 4 - 5 | 1268 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |