Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169481.2 | NP_001162952.1 | 6 - 7 | 2117 | 71 | |
NM_001273449.1 | NP_001260378.1 | 5 - 6 | 2070 | 71 | ||
NM_001273450.1 | NP_001260379.1 | 4 - 5 | 1767 | 71 | ||
NM_057214.5 | NP_476562.1 | 6 - 7 | 2169 | 71 | ||
NM_057215.5 | NP_476563.1 | 6 - 7 | 2165 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |