Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297886.1 | NP_001284815.1 | 3 - 4 | 3404 | 127 | |
NM_057243.4 | NP_476591.1 | 3 - 4 | 3404 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |