Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057280.4 | NP_476628.2 | 2 - 3 | 1550 | 76 | |
GenBank mRNA | AY119654.1 | - | 2 - 3 | 1543 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |