Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mle | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057293.4 | NP_476641.1 | 1 - 2 | 155 | 68 | |
GenBank mRNA | BT010267.1 | - | 1 - 2 | 151 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |