Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103683.2 | NP_001097153.1 | 5 - 6 | 3787 | 95 | |
NM_001298947.1 | NP_001285876.1 | 3 - 4 | 2119 | 95 | ||
NM_057311.5 | NP_476659.1 | 3 - 4 | 2557 | 95 | ||
GenBank mRNA | AY118387.1 | - | 3 - 4 | 2118 | 95 | |
M81957.1 | - | 3 - 4 | 2118 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |