Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: peb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014722.2 | NP_001014722.1 | 2 - 3 | 1388 | 58 | |
NM_057326.5 | NP_476674.3 | 2 - 3 | 1393 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |