Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp61F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057339.5 | NP_476687.1 | 1 - 2 | 226 | 72 | |
NM_057340.4 | NP_476688.1 | 1 - 2 | 226 | 72 | ||
GenBank mRNA | BT133407.1 | - | 1 - 2 | 156 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |