Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057353.4 | NP_476701.1 | 1 - 2 | 634 | 80 | |
GenBank mRNA | BT044485.1 | - | 1 - 2 | 634 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |