Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057354.3 | NP_476702.1 | 5 - 6 | 1532 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |